Description
Fibroblast Monoclonal antibody specifically detects Fibroblast in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | Fibroblast |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | ER-TR7 |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | reticular fibroblast |
| Host Species | Rat |
| Immunogen | Isolated C3H thymic stromal cells |
| Purification Method | Protein G purified |
| Show More |