missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin 1 Antibody (CL0337) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52918APC
This item is not returnable.
View return policy
Description
Fibulin 1 Monoclonal antibody specifically detects Fibulin 1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| Fibulin 1 | |
| Monoclonal | |
| APC | |
| FBLN, FIBL1, FIBL-1, fibulin 1, fibulin-1 | |
| This antibody was developed using a recombinant protein derived from P23142, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 2192 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0337 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction