missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin 1 Antibody (CL0337) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52918CL1
This item is not returnable.
View return policy
Description
Fibulin 1 Monoclonal antibody specifically detects Fibulin 1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| Fibulin 1 | |
| Monoclonal | |
| CoraFluor 1 | |
| FBLN, FIBL1, FIBL-1, fibulin 1, fibulin-1 | |
| This antibody was developed using a recombinant protein derived from P23142, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 2192 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0337 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur