Learn More
Invitrogen™ Fibulin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551665
Description
Immunogen sequence: TLGSFHCYKA LTCEPGYALK DGECEDVDEC AMGTHTCQPG FLCQNTKGSF YCQARQRCMD GFLQDPEGNC VDINECTSLS EPCRPGFSCI NTVGSYTCQR NPLICARGYH ASDDGTKCVD VNE Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
This gene encodes an extracellular matrix protein, which belongs to the fibulin family. This protein binds various extracellular ligands and calcium. It may play a role during organ development, in particular, during the differentiation of heart, skeletal and neuronal structures. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| Fibulin 2 | |
| Polyclonal | |
| Unconjugated | |
| FBLN2 | |
| 5730577E14Rik; FBLN2; FIBL2; FIBL-2; fibulin 2; Fibulin2; fibulin-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2199 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P98095 | |
| FBLN2 | |
| Recombinant protein corresponding to Human Fibulin 2. Recombinant protein control fragment (Product #RP-102097). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.