Learn More
CELLPATH FILOSLIDE 20 SLIDE TRAY
Product Code. 30226324
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Color:
Black
Blue
Green
Red
White
Yellow
missing translation for 'unitSize'
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
30226324 Blue Pack of 10
30226321 Black Pack of 10
30226322 White Pack of 10
30226323 Green Pack of 10
30226325 Yellow Pack of 10
30226326 Red Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
Product Code. 30226324 missing translation for 'mfr' CELLPATH missing translation for 'supplierNo' WEX200601B
Add to basket
Add to basket
The FiloSlide 20 slide trays feature a useful 360° cover flap, which neatly folds back on itself to save working space. An index reference is included on the flap and a recessed well on the inside of this flap prevents damp slides from sticking to the board.
  • The FiloSlide 20 tray, which measures 335 mm x 204 mm has a single column of 20 wells.
  • Each slide has its own easy release compartment which is individually numbered.
  • Constructed from solid board and available in six colours including black, white, yellow, blue, green and red.
  • This allows laboratories to colour code different types of workload or different applications.
  • Key Benefits:
  • 360° cover flap saves working space requirement
  • Range of colours available to assist in team resource allocation
  • Thumb grooves for easy removal of slides
  • Robust, allowing multiple use and cost savings
Specifications
Type Slide Holder
Slide Capacity 20
Color Blue