missing translation for 'onlineSavingsMsg'
Learn More

Fipronil Detrifluoromethylsulfinyl, TRC

Product Code. 30137547
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137547 1 g 1g
30136649 500 mg 500mg
30010329 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137547 Supplier Toronto Research Chemicals Supplier No. TRCF3422201G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120068-79-3
Molecular Formula C11 H5 Cl2 F3 N4
Molecular Weight (g/mol) 321.09
Synonym 1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-3-cyano-5-aminopyrazole, 5-Amino-1-(2,6-dichloro-4-trifluoromethylphenyl)-3-cyanopyrazole, 5-Amino-3-cyano-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-1H-pyrazole, 5-Amino-3-cyano-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]pyrazole;
IUPAC Name 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]pyrazole-3-carbonitrile
SMILES Nc1cc(nn1c2c(Cl)cc(cc2Cl)C(F)(F)F)C#N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H5Cl2F3N4/c12-7-1-5(11(14,15)16)2-8(13)10(7)20-9(18)3-6(4-17)19-20/h1-3H,18H2
Formula Weight 319.9843
Chemical Name or Material Fipronil Detrifluoromethylsulfinyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.