Learn More
Esculape Laboratoire™ First Aid Kit Refill
Product Code. 11718403
Change view
Click to view available options
Sufficient For:
10 Persons
8 People
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
11718403 8 People Each
11758403 10 Persons Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11718403 Supplier Esculape Laboratoire™ Supplier No. 805 05 05
  • ONLINE EXCLUSIVE PRICE
    57.75€
    31.57€ / Pack of 1
    Save 26.18€
    45% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Refill kit for Esculape Laboratoire first aid kit
  • Refill kit
  • For Esculape Laboratoire first aid kit
Specifications
For Use With (Application) First Aid Kit
Product Type Refill Kit
Sufficient For 8 People