Learn More
Esculape Laboratoire™ First Aid Kit Refill
Product Code. 11758403
Change view
Click to view available options
Sufficient For:
10 Persons
8 People
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
11758403 10 Persons Each
11718403 8 People Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11758403 Supplier Esculape Laboratoire™ Supplier No. 805 09 09
  • ONLINE EXCLUSIVE PRICE
    84.00€
    46.26€ / Pack of 1
    Save 37.74€
    44% Off
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
Refill kit for Esculape Laboratoire first aid kit
  • Refill kit
  • For Esculape Laboratoire first aid kit
Specifications
For Use With (Application) First Aid Kit
Product Type Refill Kit
Sufficient For 10 Persons