Learn More
Esculape Laboratoire™ First Aid Kits
Product Code. 11728383
Change view
Click to view available options
Height (Metric):
8 cm
11 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
11728383 8 cm Each
11738383 11 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11728383 Supplier Esculape Laboratoire™ Supplier No. 721 02 05
Estimated Shipment: 18-08-2026
Specifications
Depth (Metric) 21 cm
Length (Metric) 23 cm
Product Type First Aid Kit
Color Red
Includes 2 Gas band 3m x 7cm, 2 Extensible web 3m x 7cm, 3 Anti-compress shots, 10 Sterile compresses 20 x 20, Doigtier, Guide first aid, 6 number of clips, scissors, Pair of Vinyl gloves, Sterile pressure dressing 2.5m x 10cm, sterile pressure dressing 2.5 x 6cm,
Quantity 1 Ea.
Height (Metric) 8 cm