Learn More
Fisher BioReagents™ EZ-Run™ Rec Protein Ladder
Description
- Ladder is ideal for precise molecular weight determination of polypeptides in denaturing PAGE and Western blots
- Each protein contains an integral Strep-tag™ II sequence1 and may be detected on Western blots using Strep-Tactin™-AP conjugate2 or an antibody against Strep-tag II
- 50kDa protein band has greater intensity than other bands to serve as a reference
- Supplied in gel loading buffer
Specifications
Specifications
| Form | Liquid |
| Formulation | Supplied in gel-loading buffer |
| For Use With (Application) | Molecular weight determination of polypeptides analyzed in various Gel system |
| Kit Contents | <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet |
| Quantity | 500 μL, 100 Loadings |
| Range | 10 to 200 kd |
| Storage Requirements | −20°C |
| Sufficient For | 100 Loadings |
| Product Type | Protein Molecular Weight Marker |