Learn More
Fisherbrand™ 150 and 850 Hand Held Homogenizer Accessory, Stainless Steel Probes
GREENER_CHOICE
Product Code. 15505829 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Bottom Type:
Flat
Sawtooth
Sawtooth, wide window
Volume Range (Metric):
0.2 to 5 mL
0.25 to 30 mL
1.5 to 100 mL
Diameter (Metric) Probe:
5 mm
7 mm
10 mm
Particle Size:
Course
Fine
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bottom Type Volume Range (Metric) Diameter (Metric) Probe Particle Size Unit Size
15505829 Sawtooth, wide window 1.5 to 100 mL 10 mm Course Each
15545819 Flat 0.2 to 5 mL 5 mm Fine Each
15555819 Sawtooth 0.25 to 30 mL 7 mm Fine Each
15565819 Sawtooth, wide window 0.25 to 30 mL 7 mm Course Each
15595819 Sawtooth 1.5 to 100 mL 10 mm Fine Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Warning: Stainles Steel bearings cannot be autoclaved.
Product Code. 15505829
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
Warning: Stainles Steel bearings cannot be autoclaved.
Choice of flat or saw tooth tips for use with hand-held homogenizers.
  • Provides end user flexibility to process samples
  • Designed for long life and are easy to disassemble for cleaning
  • Stator design allows fine to medium samples to pass through the processing windows
  • Comes with PTFE  upper and lower bearings
TRUSTED_SUSTAINABILITY
Specifications
Product Type Probe
Particle Size Course
Volume Range (Metric) 1.5 to 100 mL
Diameter (Metric) Probe 10 mm
Length (English) Probe 4.5 in.
Length (Metric) Probe 115 mm
Material Stainless Steel
Bottom Type Sawtooth, wide window
For Use With (Equipment) 150 Hand Held Homogenizer and 850 Homogenizers