Learn More
Fisherbrand™ Traceable™ Digital Thermometer with Stainless-Steel Stem, 0.25 in. LCD Screen, and Protective Guard
Product Code. 11709715 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +150°C
-50°C to +250°C
-50°C to +280°C
-50°C to +300°C
Temperature Range (English):
-58°F to +302°F
-58°F to +482°F
-58°F to +536°F
-58°F to +572°F
Accuracy:
±0.2°C
±0.4°C
±0.5°C
±1°C
±1.5°C
Length (Metric) Probe:
8.9 cm
20 cm
20.32 cm
28.87 cm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Temperature Range (English) Accuracy Length (Metric) Probe unitSize
11709715 -50°C to +300°C -58°F to +572°F ±0.4°C 8.9 cm Each
11759715 -50°C to +280°C -58°F to +536°F ±0.4°C 20 cm Each
11719715 -50°C to +150°C -58°F to +302°F ±0.2°C 20 cm Each
11749715 -50°C to +280°C -58°F to +536°F ±1°C 20.32 cm Each
11729715 -50°C to +250°C -58°F to +482°F ±1°C 28.87 cm Each
11739715 -50°C to +300°C -58°F to +572°F ±0.5°C 28.87 cm Each
11789705 -50°C to +300°C -58°F to +572°F ±1.5°C 8.9 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Récupel-bijdrage uitgesloten
Product Code. 11709715
Récupel-bijdrage uitgesloten
Easy-to-read handheld Traceable™ Digital Thermometer.
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
Certifications
Multi-point calibration on individually-numbered Traceable Certificate assures accuracy from ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA; indicates traceability to NIST.
Specifications
Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.14 in.
Diameter (Metric) Probe 3.56 mm
For Use With (Application) Measuring temperature in soil, cuvettes, test tubes, food products, incubators and petroleum hydrocarbons
Includes Traceable certificate, Stainless steel probe, Protective case, Silver oxide battery
Length (English) Probe 3.5 in.
Length (Metric) 177 mm
Length (Metric) Probe 8.9 cm
Material Plastic
Resolution 0.1°C
Weight (English) 1 oz.
Weight (Metric) 28 g
Product Type Digital Thermometer
Length (English) 7 in.
Max. Temperature (English) 572°F
Max. Temperature (Metric) 300°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +572°F
Temperature Range (Metric) -50°C to +300°C
Calibrated Yes
Accuracy ±0.4°C
Display Digital LCD
Width (Metric) 8 cm