Learn More
Fisherbrand™ DNA Sequencing Systems: Spacer for Model FB-SEQ-3545, Spacer
Product Code. 11937515 Shop All Thermo Scientific Products
Change view
Click to view available options
Height (Metric):
0.04 cm
Thickness (Metric) Teeth:
0.4 mm
Unit Size:
Pack of 2
Pack of 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Thickness (Metric) Teeth Unit Size
11937515 0.04 cm 0.4 mm Pack of 2
11525265 0.04 cm 0.4 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11937515 Supplier Fisherbrand™ Supplier No. S2SSA4
  • ONLINE EXCLUSIVE PRICE
    42.90€
    40.60€ / Pack of 2
    Save 2.30€
    5% Off
Estimated Shipment: 18-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
DNA Sequencing Systems Spacers for model FB-SEQ-3545
For use with model FB-SEQ-3545
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Sequencing gel casting systems
Height (English) Footprint 4.8 in.
Height (Metric) 0.04 cm
Height (Metric) Footprint 12.2 cm
Length (Metric) 45 cm
Product Type Spacer Set
Thickness (Metric) Teeth 0.4 mm
Width (Metric) 1 cm
Width (Metric) Footprint 21 cm