Learn More
Fisherbrand™ Comb for use with FB-VE10-1 Vertical Electrophoresis System
Product Code. 12464657 Shop All Thermo Scientific Products
Change view
Click to view available options
Thickness (Metric) Teeth:
0.05 cm
0.08 cm
0.15 cm
0.77 cm
Teeth Count:
6
8
10
12
15
20
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Teeth Teeth Count Unit Size
12464657 0.05 cm 12 Each
12404667 0.77 cm 8 Each
11983015 0.15 cm 6 Each
11973015 0.08 cm 6 Each
11993015 0.08 cm 8 Each
11983025 0.15 cm 8 Each
11933015 0.08 cm 10 Each
12404657 0.05 cm 10 Each
11953015 0.08 cm 12 Each
12484657 0.05 cm 20 Each
12474657 0.05 cm 15 Each
11963015 0.15 cm 12 Each
12494657 0.05 cm 6 Each
11943015 0.15 cm 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 12464657 Supplier Fisherbrand™ Supplier No. MP12A
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P8 Series Electrophoresis System
Type Comb
Gel Dimensions 10 x 10 cm
Teeth Count 12
Thickness (Metric) Teeth 0.05 cm
Volume (Metric) Well 34 μL
Width (English) Teeth 0.16 in.
Width (Metric) Teeth 0.43 cm