Learn More
Fisherbrand™ Aluminum Weighing Dishes
Product Code. 11799408 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
20 mL
42 mL
65 mL
70 mL
150 mL
Depth (Metric):
13 mm
16 mm
17 mm
17.3 mm
20 mm
Diameter (Metric) Inner Base:
44 mm
51 mm
68.2 mm
69 mm
96 mm
Shape:
Flanged Sides
Fluted Sides
Unit Size:
Pack of 100
Pack of 1000
Pack of 144
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Depth (Metric) Diameter (Metric) Inner Base Shape unitSize
11799408 65 mL 16 mm 69 mm Fluted Sides Pack of 1000
10297593 42 mL 17 mm 51 mm Fluted Sides Pack of 144
11789408 20 mL 13 mm 44 mm Fluted Sides Pack of 1000
11709418 70 mL 17.3 mm 68.2 mm Fluted Sides Pack of 1000
11719418 150 mL 20 mm 96 mm Flanged Sides Pack of 1000
11518382 150 mL 20 mm 96 mm Flanged Sides Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11799408
Spotlight Offers
Use promo code 28036
View all promotions
In Stock
Add to basket
Edge
Add to basket
Aluminum dishes for weighing and dispensing
Choose Fisherbrand™ Aluminum Weight Dishes for weighing and dispensing.
With Fluted sides
  • Also can serve as evaporating dishes or dust covers
  • Lightweight, yet sturdy
  • Convenient finger-grip handle can be etched prior to weighing for identification
With Smooth sides
  • Lightweight dishes with flanged rims
  • Slightly sloping sides facilitate stacking
  • Especially useful for materials that cannot easily be cleaned from dish surface
Specifications
Bottom Shape Flat
Capacity (Metric) 65 mL
Material Aluminum
Shape Fluted Sides
Type Weighing Dish
For Use With (Application) For weighing and dispensing
Depth (Metric) 16 mm
Diameter (Metric) Inner Base 69 mm
Diameter (Metric) Inner Top 77 mm
Diameter (Metric) 69 mm (Inner Base), 77 mm (Inner Top)