missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KPNA3 Polyclonal Antibody, Biotin

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142095
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
Specifications
| KPNA3 | |
| Polyclonal | |
| Biotin | |
| KPNA3 | |
| hSRP1; importin; importin alpha 4; importin alpha Q2; importin alpha-3; importin subunit alpha-3; importin subunit alpha-4; importin-alpha-Q2; IPOA4; karyopherin (importin) alpha 3; karyopherin alpha 3; karyopherin alpha 3 (importin alpha 4); karyopherin subunit alpha 3; karyopherin subunit alpha-3; Kpna3; predicted role in nuclear import; qip2; RP11-432M24.3; SRP1; SRP1gamma; SRP1-gamma; SRP4 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 16648, 361055, 3839 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O00505, O35344 | |
| KPNA3 | |
| Peptide with sequence CDPTANLQTKEFNF (aa 509-521). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction