Learn More
Fisherbrand™ Fisherbrand™ 51 Expansion Glass Shell Vials with Plug Style Closures
Product Code. 10016474 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Volume (Metric):
3.7 mL
7.4 mL
25.8 mL
Height (Metric):
45 mm
51 mm
65 mm
Volume (English):
1 dr.
2 dr.
7 dr.
Diameter (Metric) Outer:
15 mm
19 mm
29 mm
Unit Size:
Pack of 144
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Height (Metric) Volume (English) Diameter (Metric) Outer Unit Size
10016474 7.4 mL 51 mm 2 dr. 19 mm Pack of 144
12634235 25.8 mL 65 mm 7 dr. 29 mm Pack of 144
12851861 3.7 mL 45 mm 1 dr. 15 mm Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10016474
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Fisherbrand™ 51 Expansion Glass Shell Vials with Plug Style Closures are straight-sided TITESEAL vials ideal for storing dry contents. Made from 51 expansion borosilicate glass meeting USP Type I and ASTM E438, Type I, Class B requirements, with open-bottom plug style plastic closures.
Straight-sided shell vials are ideal for dry contents.
  • TITESEAL™ vials designed for dry contents
  • Straight-sided shell vial design with plain tops
  • Open-bottom plug style plastic closures, unattached
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
  • Available in multiple dram sizes to suit a range of storage needs
  • Supplied in bulk pack quantities
Order Info
Vials without closures use corks (sold separately)
Specifications
Color Clear
Diameter (Metric) Outer 19 mm
Material Borosilicate Glass
Pre-slit No
Sterility Non-sterile
Disposable Single-use
Volume (Metric) 7.4 mL
For Use With (Application) Ideal for storing dry products
Volume (English) 2 dr.
Product Line TITESEAL
Volume (Metric) Total 7.4 mL
Closure Material Plastic
Closure Type Plug
Quantity 1152/Cs.
Marking Spot No
Autoclavable Autoclavable
Certifications/Compliance ASTM E438
Height (Metric) 51 mm
Product Type Shell Vial