Learn More
Fisherbrand™ Type 316 Stainless-Steel Pressure Vessel
Description
- For dispensing and processing liquids.
†Top opening.
Specifications
Specifications
| Material | Stainless Steel (316) |
| Type | Pressure Vessel |
| Volume (Metric) | 3.8 L |
| Height (English) | 8.5 in. |
| Height (Metric) | 21.6 cm |