Learn More
Fisherbrand™ Urine Sedimentation System: Bulb Decanting Pipette
Product Code. 11947075
Encompass
Change view
Click to view available options
Quantity:
500 Case
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11947075 500 Case Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11947075 Supplier Fisherbrand™ Supplier No. 14375560
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
The Fisherbrand™ Urine Sedimentation System is designed to produce the same results as the most popular style urine sedimentation system using the same test procedure
Decanting pipet allows supernatant to be decanted off; leaves 1mL of urine-containing sediment in the bottom of the tube.
Specifications
Type Bulb Decanting Pipette
For Use With (Application) Designed to produce the same results as the most popular style Urine sedimentation system using the same test procedure
Quantity 500 Case