missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FITC Monoclonal Antibody (931845)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561981
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs ultraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.
Specifications
| FITC | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Mouse | |
| Protein A/G | |
| RUO | |
| All species | |
| Antibody | |
| IgG2a |
| Immunocytochemistry | |
| 931845 | |
| Unconjugated | |
| Fluorescein | |
| Fluorescein Isothiocyanate (FITC). | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction