Learn More
VELP Scientifica FIWE Advance Automatic Fiber Analyzer
Product Code. 17851849 Shop All Velp Products
Change view
Click to view available options
Voltage:
115/230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
17851849 115/230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17851849 Supplier VELP Scientifica Supplier No. F30500500
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Fully automatic analyzer for crude and detergent fiber determination according to official reference methods requiring minimum operator time with unique user interface and cloud connectivity.
COMPLIANCE AND CONSISTENCY
  • The crucible method is adherent to official standards (ISO and AOAC) ensuring reliability and efficiency.
  • Crude Fiber determination and Detergent Fiber determination of NDF, ADF and ADL.
  • Fiber residues stay in crucibles during all steps of the analysis eliminating the risk of errors and ensuring consistent results.
LOAD THE SAMPLES AND GO
  • The FIWE Advance heats and doses the required reagents precisely to analyze up to 6 samples.
  • The VELP Dispensing Nozzle drives the reagents precisely into each column avoiding user contact with hot chemicals and fumes.
  • FIWE Advance automates digestion, filtration and washing steps without any operator intervention.
SAVING TIME AND MONEY
  • The operator time required is as little as 2 minutes.
  • The FIWE Advance treats each sample individually and takes care of each step of the analysis allowing for overnight work.
  • The FIWE Advance automatic processes free up valuable time enhancing lab productivity and saving money.
COMPLETE SAFETY
  • All the reagents required are contained in dedicated glass tanks and bottles located inside the instrument.
  • The FIWE Advance pre-heats, dispenses, and collects hot chemicals automatically so the risk of contact with the operator is eliminated.
SMART AND EASY TO USE
  • The 7 in. color touch screen offers simple and intuitive user interface designed to simplify operations.
  • Choose from preset methods or customize your own, mark as your favorites for quick access.
  • Optional balance connection and barcode scanner are available to simplify analysis preparation.
Specifications
Type Automatic Fiber Analyzer
Voltage 115/230 V