Learn More
Trajan™ Fixed-Needle Autosampler Syringes for Thermo Scientific Instruments
Product Code. 16672572
Change view
Click to view available options
Needle Gauge:
22 G
23 G
25 G
26 G
Length (English) Needle:
1.96 in.
2 in.
3.14 in.
Model:
10F-BT-5/0.5C
10F-BT-8/0.47C
10F-BT-8/0.63C
10F-BT-8/22C
10F-BT-GT-5/0.47C
10F-BT-GT-5/0.63C
5F-BT-5/0.47C
Quantity:
1 Ea.
6 Pk.
Ea.
Unit Size:
Each
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Length (English) Needle Model Quantity Unit Size
16672572 22 G 3.14 in. 10F-BT-8/22C Ea. Each
15258443 26 G 1.96 in. 5F-BT-5/0.47C Ea. Each
15228463 26 G 1.96 in. 10F-BT-GT-5/0.47C Ea. Each
15298463 26 G 3.14 in. 10F-BT-8/0.47C 1 Ea. Each
15278463 23 G 2 in. 10F-BT-GT-5/0.63C 1 Ea. Each
15208463 25 G 1.96 in. 10F-BT-5/0.5C Ea. Each
15288463 23 G 3.14 in. 10F-BT-8/0.63C Ea. Each
15218463 26 G 1.96 in. 10F-BT-GT-5/0.47C 6 Pk. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 16672572 Supplier Trajan™ Supplier No. 0029741
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For reduced non specific sample interaction and increased analyte recovery
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Needle Gauge 22 G
Diameter (English) Outer Needle 0.02 in.
Diameter (Metric) Outer Needle 0.72 mm
For Use With (Application) Gas chromatography
Length (English) Needle 3.14 in.
Product Type Autosampler Syringe
Model 10F-BT-8/22C
Quantity Ea.
Volume (Metric) 10 μL
Needle Point Style Cone
For Use With (Equipment) Thermo AS2000
Connection Type Fixed