Learn More
Thermo Scientific Fixed Price Preventative Maintenance for Cold Storage, Freezer, Refrigerator, ULT AND Arctiko CRYO -150°C Compressor-Based Freezer
Product Code. 32176298 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
ULT Freezer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
32176298 ULT Freezer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32176298 Supplier Thermo Scientific Supplier No. FPPMULTMVS
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Scope covers compressor-based refrigerators, freezers, ULT and Arctiko CRYO -150°C equipment where LN2 backup is secondary to the primary cooling system.
  • Not suitable for liquid nitrogen Dewar-type cryopreservation systems
  • Parts, consumables, corrective repairs and primary LN2 cryopreservation systems are excluded unless separately quoted
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ULT Freezer
Product Type Preventive Maintenance