Learn More
Thermo Scientific Fixed Price Preventative Maintenance for Fisherbrand Biosafety Cabinets
Product Code. 30503419
Change view
Click to view available options
For Use With (Equipment):
Biosafety Cabinets
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
30503419 Biosafety Cabinets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30503419 Supplier Thermo Scientific Supplier No. FPPMBSCFIS
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fixed-cost planned maintenance for Fisherbrand biosafety cabinets, including KI Discus test where required by national regulations.
  • In accordance with the European Standard for microbiological safety cabinets (CEN BS EN12469:2000, Table 5), the KI Discus test is identified as an optional test for operator/personal protection.
  • Applies to both commissioning and annual routine maintenance testing.
  • National regulations must be followed in all cases.
  • Some national regulations, such as those from the HSE (UK), recommend performing the KI Discus test at least annually.
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) Biosafety Cabinets
    Product Type Preventive Maintenance