Learn More
FKBPL Monoclonal Antibody (OTI4G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502051
Description
The protein encoded by this gene has similarity to the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The encoded protein is thought to have a potential role in the induced radioresistance. Also it appears to have some involvement in the control of the cell cycle.Specifications
| FKBPL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UIM3 | |
| FKBPL | |
| Full length human recombit protein of human FKBPL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI4G1 | |
| Unconjugated | |
| FKBPL | |
| DIR1, FKBP4, NG7, WISP39 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 63943 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.