Learn More
FKBPL Monoclonal Antibody (OTI4G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502051S
Description
FKBPL Monoclonal antibody specifically detects FKBPL in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| FKBPL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y3Z3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 25939 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI4G1 | |
| Unconjugated | |
| SAMHD1 | |
| DIR1; FK506 binding protein like; FK506 binding protein-like; FK506-binding protein like; FK506-binding protein-like; FKBP4; FKBPL; immunophilin-like protein NG7; NG7; peptidyl-prolyl cis-trans isomerase; Ppiase-X; WAF-1/CIP1 stabilizing protein 39; WISp39 | |
| Full length human recombinant protein of human SAMHD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.