Learn More
Glassco Laboratory Equipments Flat Bottom Glass Flask
Product Code. 31937667
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 10
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31937667 50 mL Pack of 10
31937684 100 mL Pack of 10
31937690 250 mL Pack of 10
31937691 500 mL Pack of 10
31937655 1000 mL Pack of 10
31937656 2000 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31937667 Supplier Glassco Laboratory Equipments Supplier No. EU058.202.02A
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Flat bottom glass flask with a joint for general laboratory use.
  • Flat bottom glass flasks with an NS 29/32 joint
  • Available in 50 mL, 100 mL, 250 mL, 500 mL, 1000 mL, and 2000 mL capacities
  • Made from 3.3 borosilicate glass
  • Complies with DIN/ISO 4797 and USP standards
  • Features white printing
  • Chemical resistant
Specifications
Capacity (Metric) 50 mL
Material Borosilicate Glass (3.3)
Product Type Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Joint
Sterility Non-sterile
Bottom Shape Flat
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.