missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAP1 Antibody (OTI1H8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01707
This item is not returnable.
View return policy
Description
TRAP1 Monoclonal specifically detects TRAP1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TRAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| TRAP1 | |
| Full length human recombinant protein of human TRAP1(NP_057376) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1H8 | |
| Unconjugated | |
| HSP 75, HSP75TNFR-associated protein 1, HSP90Lheat shock protein 75 kDa, mitochondrial, TNF receptor-associated protein 1, TRAP-1, tumor necrosis factor type 1 receptor associated protein, Tumor necrosis factor type 1 receptor-associated protein | |
| Mouse | |
| 79.9 kDa | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 10131 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction