Learn More
Esculape Laboratoire™ Flexible Special Rescue Kit
Product Code. 11739112
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11739112 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11739112 Supplier Esculape Laboratoire™ Supplier No. 7180405
Estimated Shipment: 24-07-2026
Specifications
Depth (Metric) 4 cm
Length (Metric) 9 cm
Product Type Flexible Special Rescue Kit
Includes 1 survival blanket, 1 face shield mouth to mouth, 2 sterile compresses 20 x 20cm, 1 compression bandage 6 x 250cm, 2 to the disinfecting wipes, 1 pair of Vinyl gloves, first aid kit flexible
Quantity 1 Ea.
Height (Metric) 13 cm