Learn More
Molecular Devices FlexStation™ 3 Microplate Reader without Pipettor Head
Product Code. 31935892 Shop All Molecular Devices Products
Change view
Click to view available options
Detector Type:
Photomultiplier Tube (PMT)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detector Type Unit Size
31935892 Photomultiplier Tube (PMT) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 31935892 Supplier Molecular Devices Supplier No. FLEX3
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Perform flexible microplate‐based assays with integrated detection capabilities. Supports multi‐mode workflows for biochemical and cell‐based applications.
  • Designed for multi-mode detection workflows in microplate-based assays
  • Supports biochemical and cell-based assay applications
  • Enables flexible assay setup with configurable detection parameters
  • Supports microplate-based workflows across multiple life science applications
  • Designed for integration with automated liquid handling using optional pipettor head
  • Integrated with SoftMax Pro Software for data acquisition and analysis
  • Supports scalable assay workflows for research environments
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microplate Reader
Detector Type Photomultiplier Tube (PMT)
Interface PC Control, Internal Software
Light Source Xenon Flash Lamp
Measurement Technology Fluorescence Intensity (Top and Bottom), Luminescence (Top), Absorbance (Top)
Plate Type 96 and 384-well Plates
Reading Options Endpoint, Kinetic, Spectra
Shaking Linear
Wavelength Selection Monochromator