missing translation for 'onlineSavingsMsg'
Learn More

Floctafenine, TRC

Product Code. 30134791
Change view
Click to view available options
Quantity:
10 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134791 200 mg 200mg
30137782 100 mg 100mg
17944375 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134791 Supplier Toronto Research Chemicals Supplier No. TRCF401700200MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23779-99-9
Molecular Formula C20 H17 F3 N2 O4
Molecular Weight (g/mol) 406.36
Synonym Benzoic acid, 2-[[8-(trifluoromethyl)-4-quinolinyl]amino]-, 2,3-dihydroxypropyl ester, Anthranilic acid, N-[8-(trifluoromethyl)-4-quinolyl]-, 2,3-dihydroxypropyl ester (8CI), Glycerol, 1-[N-[8-(trifluoromethyl)-4-quinolyl]anthranilate] (8CI), Diralgan, Floctafenin, Floctafenine, Idalon, Idarac, Novodolan, R 4138, R 4318, RU 15750
IUPAC Name 2,3-dihydroxypropyl 2-[[8-(trifluoromethyl)quinolin-4-yl]amino]benzoate
SMILES OCC(O)COC(=O)c1ccccc1Nc2ccnc3c(cccc23)C(F)(F)F

Specifications

Quantity 200 mg
InChI Formula InChI=1S/C20H17F3N2O4/c21-20(22,23)15-6-3-5-13-17(8-9-24-18(13)15)25-16-7-2-1-4-14(16)19(28)29-11-12(27)10-26/h1-9,12,26-27H,10-11H2,(H,24,25)
Formula Weight 406.114
Chemical Name or Material Floctafenine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.