Learn More
Brady™ Floor Stand
Product Code. 15193289 Shop All Brady Products
Change view
Click to view available options
Color:
Black, Red on Yellow
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15193289 Black, Red on Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15193289 Supplier Brady™ Supplier No. 241971
  • ONLINE EXCLUSIVE PRICE
    78.25€
    70.71€ / Pack of 1
    Save 7.54€
    9% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use the Floor Stand for site identification.
  • Economy duty floor stand.
Specifications
Alternate Part Number ECONOMY DUTY FLOOR STND VERBODEN TOEGANG
Displayed Text VERBODEN TOEGANG
Height (English) 25.98 in.
Height (Metric) 660 mm
Shape Rectangle
Color Black, Red on Yellow
Material Polyester
Type Floor Stand