Learn More
Paragon Scientific Flow Cup Viscosity Standards, Paragon Scientific™
Product Code. 16060172
Change view
Click to view available options
Viscosity:
1044 mPa.s (20°C)
135.9 mPa.s (20°C)
17.78 mPa.s (20 °C)
1882 mPa.s (20 °C)
281.4 mPa.s (20°C)
37.22 mPa.s (20°C)
557.7 mPa.s (20°C)
72.9 mPa.s (20°C)
8.945 mPa.s (20°C)
Unit Size:
500mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Unit Size
16060172 135.9 mPa.s (20°C) 500mL
16050172 8.945 mPa.s (20°C) 500mL
15929959 17.78 mPa.s (20 °C) 500mL
16030172 37.22 mPa.s (20°C) 500mL
16040172 72.9 mPa.s (20°C) 500mL
15939959 281.4 mPa.s (20°C) 500mL
15949959 557.7 mPa.s (20°C) 500mL
15959959 1044 mPa.s (20°C) 500mL
15969959 1882 mPa.s (20 °C) 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 16060172 Supplier Paragon Scientific Supplier No. C60
  • ONLINE EXCLUSIVE PRICE
    96.20€
    74.36€ / 500mL
    Save 21.84€
    22% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
These standards are especially designed for use in DIN, Ford, ISO, Shell and Zahn flow cups. Certified at 20°C and 25°C, each value is tested to ASTM D2162 for viscosity measurement and ASTM D1480 for Density measurement, certified to the International Standards BS EN ISO / IEC 17025 and ISO 17034.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Manufactured in accordance with ASTM D2162 the primary method for viscosity
  • Density g/mL in accordance with ASTM D1480
  • Density, dynamic viscosity and kinematic viscosity quoted at all temperatures
  • Ensures full compliance to ASTM & IP test method protocol
  • Full Traceability to National Standards
  • 2 year shelf life
Specifications
Viscosity 135.9 mPa.s (20°C)
Quantity 500 mL
Nominal values quoted