missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluconazole-d4 (bismethylene-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Fluconazole-D4 |
| CAS | 1124197-58-5 |
| Molecular Formula | C13 2H4 H8 F2 N6 O |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-16-6-18-20)5-21-9-17-7-19-21/h1-3,6-9,22 H,4-5H2/i4D2,5D2 |
| SMILES | [2 H]C([2 H])(n1cncn1)C(O)(c2ccc(F)cc2F)C([2 H])([2 H])n3cncn3 |
| IUPAC Name | 1,1,3,3-tetradeuterio-2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol |
| Molecular Weight (g/mol) | 310.3 |
| Formula Weight | 310.1292 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction