missing translation for 'onlineSavingsMsg'
Learn More

Flunarizine EP Impurity A, Mikromol™

Product Code. 18060980
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18060980 25 mg 25mg
18012980 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18060980 Supplier Mikromol Supplier No. MM0175.040025
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Flunarizine EP Impurity A
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[Bis(4-fluorophenyl)methyl]piperazine
CAS 27469-60-9
Analyte or Component Names 1-[Bis(4-fluorophenyl)methyl]piperazine
Shipping Condition Room Temperature
Molecular Formula C17 H18 F2 N2
Certifications/Compliance ISO 17025
Synonym Flunarizine Dihydrochloride Imp. A (EP), Flunarizine Imp. A (EP), 1-[Bis(4-fluorophenyl)methyl]piperazine, Piperazine, 1-[bis(4-fluorophenyl)methyl]-, Piperazine, 1-[bis(p-fluorophenyl)methyl]- (8CI), 1-(Di-p-fluorobenzhydryl)piperazine, 4-[Bis(4-fluorophenyl)methyl]piperazine, N-(4,4'-Difluorobenzhydryl)piperazine, N-(Bis(4-fluorophenyl)methyl)piperazine, R 19352, T 841, Flunarizine Dihydrochloride Impurity A, Flunarizine Impurity A
InChI Formula InChI=1S/C17H18F2N2/c18-15-5-1-13(2-6-15)17(21-11-9-20-10-12-21)14-3-7-16(19)8-4-14/h1-8,17,20H,9-12H2
SMILES Fc1ccc(cc1)C(N2CCNCC2)c3ccc(F)cc3
IUPAC Name 1-[bis(4-fluorophenyl)methyl]piperazine
Molecular Weight (g/mol) 288.34
Quantity 25 mg
Formula Weight 288.1438 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.