missing translation for 'onlineSavingsMsg'
Learn More

Fluocinolone, TRC

Product Code. 30135399
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135399 500 mg 500mg
30008688 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135399 Supplier Toronto Research Chemicals Supplier No. TRCF455795500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 807-38-5
Molecular Formula C21 H26 F2 O6
Molecular Weight (g/mol) 412.42
Synonym Pregna-1,4-diene-3,20-dione, 6,9-difluoro-11,16,17,21-tetrahydroxy-, (6α,11β,16α)-, Pregna-1,4-diene-3,20-dione, 6α,9-difluoro-11β,16α,17,21-tetrahydroxy- (6CI,7CI,8CI), (6α,11β,16α)-6,9-Difluoro-11,16,17,21-tetrahydroxypregna-1,4-diene-3,20-dione, 16α-Hydroxy-6α,9-difluoroprednisolone, 6α-Fluorotriamcinolone, Fluocinolone, 6α,9-Difluoro-11β,16α,17,21-tetrahydroxypregna-1,4-diene-3,20-dione
IUPAC Name (6S,8S,9R,10S,11S,13S,14S,16R,17S)-6,9-difluoro-11,16,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@]12C[C@H](O)[C@@]3(F)[C@@H](C[C@H](F)C4=CC(=O)C=C[C@]34C)[C@@H]1C[C@@H](O)[C@]2(O)C(=O)CO

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H26F2O6/c1-18-4-3-10(25)5-13(18)14(22)6-12-11-7-15(26)21(29,17(28)9-24)19(11,2)8-16(27)20(12,18)23/h3-5,11-12,14-16,24,26-27,29H,6-9H2,1-2H3/t11-,12-,14-,15+,16-,18-,19-,20-,21-/m0/s1
Formula Weight 412.1697
Percent Purity >95%
Chemical Name or Material Fluocinolone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.