missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein disodium salt, Thermo Scientific™

Fluorescein disodium salt (uranin, fluorescite, acid yellow 73, CAS # 518-45-6) is the salt of the xanthene dye commonly used for fluorescent tracing.

Brand:  Thermo Scientific Alfa Aesar J61549.36

71.02 EUR valid until 2025-12-19
BEST PRICE on Promo! Use promo code "24110" to get your promotional price.


Quantity 500g

 View more versions of this product

Product Code. 15474139

  • ONLINE EXCLUSIVE PRICE
    84.80€
    83.10€ / 500g
    Save 1.70€
    2% Off

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• Fluorescein disodium salt is a yellow-green phthalate indicator dye that is pH-sensitive

• It has a maximum absorbance of 494nm and maximum emission of 521nm

Application

• Fluorescein disodium salt can be used as a fluorescent protein labeler

• Fluorescein is also useful for the staining of cells and organelles

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

518-47-8
C20H10Na2O5
MFCD00167039
Acid Yellow 73; C.I. 45350

Specifications

518-47-8
MFCD00167039
3833041
Acid Yellow 73; C.I. 45350
376.28
C20H10Na2O5
500g
14,4159
Soluble in water at 1mg/ml
Fluorescein disodium salt
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.