Learn More
Cytiva Fluoro Plastics Fluoro Rubber Glass Superloop
Product Code. 11340122 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11340122 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11340122 Supplier Cytiva Supplier No. 19785001
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Introduce large sample volumes into a pressurised fluid LC system. Cytiva Fluoro Plastics Fluoro Rubber Glass Superloop is used together with the injection valve MV-7, INV-907, V9-Inj, or V9H-Inj and replaces a simple sample loop.
  • Superloop can be used in conjunction with 7-port valves.
  • The patented devices enable controlled loading of large samples, either in the form of multiple small injections or a single large injection.
  • They may also be used to inject pulses of a specific buffer solution.
  • Flexible: inject large or small sample volumes.
  • High reproducibility and recovery.
  • No gradient dilution with internal movable seal.
TRUSTED_SUSTAINABILITY
Specifications
Type Superloop
Material Fluoro Plastics, Fluoro Rubber, Glass
Volume (Metric) 50 mL