missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine hydrochloride, 99%, Thermo Scientific™

A selective serotonin reuptake inhibitor

Brand:  Thermo Scientific Alfa Aesar J61197.MD

Quantity 250mg

 View more versions of this product

Product Code. 15463819

  • ONLINE EXCLUSIVE PRICE
    465.00€
    418.65€ / 250mg
    Save 46.35€
    9% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).

Solubility
Soluble in dimethylsulfoxide and water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

56296-78-7
345.79
GIYXAJPCNFJEHY-UHFFFAOYSA-N
62857
CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
C17H19ClF3NO
MFCD00214288
fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride

Specifications

56296-78-7
MFCD00214288
14,4185
Soluble in dimethylsulfoxide and water.
CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
345.79
345.79
Fluoxetine hydrochloride
C17H19ClF3NO
250mg
fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
GIYXAJPCNFJEHY-UHFFFAOYSA-N
N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
62857
99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.