missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine hydrochloride, 99%, Thermo Scientific™

A selective serotonin reuptake inhibitor

150.65€ - 418.65€

Chemical Identifiers

CAS 56296-78-7
Molecular Formula C17H19ClF3NO
Molecular Weight (g/mol) 345.79
MDL Number MFCD00214288
InChI Key GIYXAJPCNFJEHY-UHFFFAOYSA-N
Synonym fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
PubChem CID 62857
IUPAC Name N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
SMILES CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15473819
SDS View Documents
Thermo Scientific Alfa Aesar
J61197.MF
50mg
ONLINE EXCLUSIVE PRICE
167.00€
150.65€ / 50mg
Save 16.35€
9% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15463819
SDS View Documents
Thermo Scientific Alfa Aesar
J61197.MD
250mg
ONLINE EXCLUSIVE PRICE
465.00€
418.65€ / 250mg
Save 46.35€
9% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).

Solubility
Soluble in dimethylsulfoxide and water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

56296-78-7
345.79
GIYXAJPCNFJEHY-UHFFFAOYSA-N
62857
CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
C17H19ClF3NO
MFCD00214288
fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride

Specifications

56296-78-7
MFCD00214288
fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
GIYXAJPCNFJEHY-UHFFFAOYSA-N
N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
62857
99%
C17H19ClF3NO
14,4185
Soluble in dimethylsulfoxide and water.
CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl
345.79
345.79
Fluoxetine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P501c

H302-H315-H318

missing translation for 'einecsNumber' : 260-101-2

missing translation for 'rtecsNumber' : UI4050000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.