missing translation for 'onlineSavingsMsg'
Learn More

Fluoxetine hydrochloride, 99%

Product Code. 15473819
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473819 50 mg 50mg
15463819 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473819 Supplier Thermo Scientific Alfa Aesar Supplier No. J61197.MF
  • ONLINE EXCLUSIVE PRICE
    167.00€
    150.65€ / 50mg
    Save 16.35€
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A selective serotonin reuptake inhibitor | CAS: 56296-78-7 | C17H19ClF3NO | 345.79 g/mol

Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. It is used to treat major depressive disorder, bulimia nervosa (an eating disorder) obsessive-compulsive disorder, panic disorder and premenstrual dysphoric disorder (PMDD).

Solubility
Soluble in dimethylsulfoxide and water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56296-78-7
Molecular Formula C17H19ClF3NO
Molecular Weight (g/mol) 345.79
MDL Number MFCD00214288
InChI Key GIYXAJPCNFJEHY-UHFFFAOYSA-N
Synonym fluoxetine hydrochloride, prozac, fluoxetine hcl, sarafem, flunirin, fluoxeren, adofen, fluctin, lovan
PubChem CID 62857
IUPAC Name N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
SMILES CNCCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F.Cl

Specifications

Melting Point 158°C to 159°C
Color White
Quantity 50 mg
Merck Index 14,4185
Solubility Information Soluble in dimethylsulfoxide and water.
Formula Weight 345.79
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Fluoxetine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.