missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Fluvastatin sodium
Description
Orally active, potent and competitive HMG-CoA reductase inhibitor (IC50 = 40 -100 nM at human liver microsomes). Inhibits vascular smooth muscle proliferation in vitro (IC50 = 70 nM) and exhibits antihypercholesterolemic and antioxidant activity in vivo.
Specifications
Specifications
| CAS | 93957-55-2 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Molecular Formula | C24H25FNNaO4 |
| Formula Weight | 433.45 |
| Purity | >98% |
| Molecular Weight (g/mol) | 433.46 |
| Chemical Name or Material | Fluvastatin sodium |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?