missing translation for 'onlineSavingsMsg'
Learn More

Fluvastatin Sodium Salt(Relative Stereochemistry), TRC

Product Code. 30012815
Change view
Click to view available options
Quantity:
25 mg
250 mg
500 mg
Unit Size:
250mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012815 500 mg 500mg
30011099 250 mg 250mg
30011703 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012815 Supplier Toronto Research Chemicals Supplier No. TRCF601250500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93957-55-2
Molecular Formula C24 H25 F N O4 . Na
Molecular Weight (g/mol) 433.45
Synonym Fluvastatin Sodium, (3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid sodium salt (1:1), [R*,S*-(E)]-(±)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid monosodium salt, (3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid monosodium salt, Almastatin, Canef, Cranoc, Fluindostatin, Fluvastatin sodium, Fractal, Lescol, Lescol XL, Lochol, Locol, Vastin, XU 62-320
IUPAC Name sodium;(E,3R,5S)-7-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]-3,5-dihydroxyhept-6-enoate
SMILES [Na+].CC(C)n1c(\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c(c2ccc(F)cc2)c3ccccc13

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C24H26FNO4.Na/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30;/h3-12,15,18-19,27-28H,13-14H2,1-2H3,(H,29,30);/q;+1/p-1/b12-11+;/t18-,19-;/m1./s1
Formula Weight 433.1665
Chemical Name or Material Fluvastatin Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.