missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-beta-cyclopropyl-D-alanine, TRC

Product Code. 30179510
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
Unit Size:
1g
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179510 500 mg 500mg
30179440 2 g 2g
30179288 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179510 Supplier Toronto Research Chemicals Supplier No. TRCF255900500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170642-29-2
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.4
SMILES OC(=O)[C@@H](CC1CC1)NC(=O)OCC2c3ccccc3c4ccccc24

Specifications

Quantity 500 mg
Shipping Condition Room Temperature
Chemical Name or Material Fmoc-beta-cyclopropyl-D-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.