missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-Cop, TRC

Product Code. 30179240
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179240 1 g 1g
30179275 250 mg 250mg
30179096 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179240 Supplier Toronto Research Chemicals Supplier No. TRCF6227751G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312965-04-1
Molecular Formula C20H19NO5
Molecular Weight (g/mol) 353.37
Synonym 4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-2-carboxylic Acid, (R/S)-Fmoc-2-carboxymorpholine, 2,4-Morpholinedicarboxylic Acid 4-(9H-Fluoren-9-ylmethyl) Ester
SMILES OC(=O)C1CN(CCO1)C(=O)OCC2c3ccccc3c4ccccc24

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H19NO5/c22-19(23)18-11-21(9-10-25-18)20(24)26-12-17-15-7-3-1-5-13(15)14-6-2-4-8-16(14)17/h1-8,17-18H,9-12H2,(H,22,23)
Formula Weight 353.1263
Chemical Name or Material Fmoc-Cop
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.