Description
- For Research Use Only
- Not for use in diagnostics procedures
Specifications
Specifications
| Content And Storage | Each package contains 10 cartridges. |
| Format | Cartridge |
| For Use With (Equipment) | 433 A Peptide Synthesizer |
| For Use With (Application) | Protein Sequencing & Peptide Synthesis |
| Product Type | Peptide Synthesis Reagents |
| Quantity | 10 Ea. |