Learn More
Applied Biosystems™ Fmoc-L-Cysteine (Trt), 10/Pk.
Product Code. 15949172 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
10 Each
Unit Size:
Pack of 10
Product Code. Quantity unitSize
15949172 10 Each Pack of 10
1 options
Product Code. 15949172
Brand: Applied Biosystems™ 410962
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
These are Amino Acid Cartridges for the ABI™ 431-433 Peptide Synthesizer.
  • For Research Use Only
  • Not for use in diagnostics procedures
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Each package contains 10 cartridges.
Format Cartridge
For Use With (Equipment) 433 A Peptide Synthesizer
For Use With (Application) Protein Sequencing & Peptide Synthesis
Product Type Peptide Synthesis Reagents
Quantity 10 Each