missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNTB Mouse anti-Human, Mouse, Clone: 1F3B10, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | FNTB |
|---|---|
| Clone | 1F3B10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16868975
|
Proteintech
66783-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16858975
|
Proteintech
66783-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF24 is a new candidate transcription factor.Specifications
| FNTB | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| FNTB, FPTB, FTase beta | |
| FNTB | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1F3B10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| P49356, Q8K2I1 | |
| 110606, 2342 | |
| FNTB Fusion Protein Ag27796 | |
| Primary | |
| -20°C | |
| FNTB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title