missing translation for 'onlineSavingsMsg'
Learn More

Folcysteine, TRC

Product Code. 30388918
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388918 1 g
30400910 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388918 Supplier Toronto Research Chemicals Supplier No. TRCF6802001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8064-47-9
Molecular Formula C19H19N7O6 . C6H9NO3S
Molecular Weight (g/mol) 616.61
Synonym N-[4-[[(2-Amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-L-Glutamic Acid Mixture With (4R)-3-Acetyl-4-thiazolidinecarboxylic Acid, N-[4-[[(2-Amino-1,4-dihydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-L-Glutamic acid Mixture With (R)-3-Acetyl-4-thiazolidinecarboxylic Acid, Ergostim, FOP, Folcysteine FOP
IUPAC Name (4-(((2-amino-4-oxo-4,8-dihydropteridin-6-yl)methyl)amino)benzoyl)-L-glutamic acid and (R)-3-acetylthiazolidine-4-carboxylic acid
SMILES O=C(N1[C@H](C(O)=O)CSC1)C.O=C2C3=NC(CNC4=CC=C(C(N[C@H](C(O)=O)CCC(O)=O)=O)C=C4)=CNC3=NC(N)=N2

Specifications

InChI Formula InChI=1S/C19H19N7O6.C6H9NO3S/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28;1-4(8)7-3-11-2-5(7)6(9)10/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30);5H,2-3H2,1H3,(H,9,10)
Quantity 1 g
Formula Weight 616.17
Shipping Condition Room Temperature
Chemical Name or Material Folcysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.