Learn More
uvex Fold-flat Mask silv-Air p 8103 FFP1
Código de producto. 17572955 Tienda Uvex Productos
Cambiar vista
Click to view available options
Filter Type: FFP1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto.
17572955 Filter Type: FFP1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17572955 Proveedor uvex N.º de proveedor 8708103
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Este artículo no se puede devolver. Vea la política de devoluciones
The gray coloring on the outside of the silv-Air p masks prevents them from looking dirty quickly and having to be disposed of prematurely by the user. By contrast, the inside is of course pure white. In addition and with all products in this family, the 3D nose geometry ensures an optimum fit, even with different face shapes.
General Features
  • Particle-filtering flat-fold mask
  • Hydrophobic material coating
  • Seamless headband for a comfortable, secure fit
  • Flexible, adjustable integrated nose clip
  • Foam layers in chin and nose area for optimally secure fit
  • Individually sealed
  • Color: silver
Protection Features
  • FFP1 respirator certified in accordance with EN 149:2001+A1:2009
  • D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
  • Not reusable
Comfort Features
  • Seamless headband for a comfortable, secure fit
  • Flexible, adjustable integrated nose clip
  • Foam padding on the chin and nose for optimally secure fit
TRUSTED_SUSTAINABILITY
Especificaciones
Certifications/Compliance (Europe) EN 149 (D marked)
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type Particulate
Material Polypropylene, Polyester
Product Type General Purpose Particulate Respirator
Size Standard
With Exhalation Valve Yes
Filter Efficiency FFP1
Sterility Non-sterile
Mostrar más Mostrar menos
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.